Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608399 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSMD3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CSMD3 - CUB and Sushi multiple domains 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052900.2 | 13484 | UTR 3 | NP_443132.3 | |||
NM_198123.1 | 13484 | UTR 3 | NP_937756.1 | |||
NM_198124.1 | 13484 | UTR 3 | NP_937757.1 | |||
XM_011516815.2 | 13484 | UTR 3 | XP_011515117.1 | |||
XM_011516816.2 | 13484 | UTR 3 | XP_011515118.1 | |||
XM_017013008.1 | 13484 | UTR 3 | XP_016868497.1 | |||
XM_017013009.1 | 13484 | UTR 3 | XP_016868498.1 | |||
XM_017013010.1 | 13484 | UTR 3 | XP_016868499.1 | |||
XM_017013011.1 | 13484 | UTR 3 | XP_016868500.1 | |||
XM_017013012.1 | 13484 | UTR 3 | XP_016868501.1 |