Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602069 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NRP1 - neuropilin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024628.2 | 4318 | Intron | NP_001019799.1 | |||
NM_001024629.2 | 4318 | Intron | NP_001019800.1 | |||
NM_001244972.1 | 4318 | UTR 3 | NP_001231901.1 | |||
NM_001244973.1 | 4318 | UTR 3 | NP_001231902.1 | |||
NM_003873.5 | 4318 | UTR 3 | NP_003864.4 | |||
XM_006717521.1 | 4318 | UTR 3 | XP_006717584.1 | |||
XM_006717522.1 | 4318 | UTR 3 | XP_006717585.1 | |||
XM_006717523.2 | 4318 | UTR 3 | XP_006717586.1 | |||
XM_006717524.1 | 4318 | UTR 3 | XP_006717587.1 | |||
XM_006717525.1 | 4318 | UTR 3 | XP_006717588.1 | |||
XM_006717526.1 | 4318 | Intron | XP_006717589.1 | |||
XM_011519755.2 | 4318 | Intron | XP_011518057.1 | |||
XM_011519756.1 | 4318 | Intron | XP_011518058.1 | |||
XM_017016865.1 | 4318 | UTR 3 | XP_016872354.1 | |||
XM_017016866.1 | 4318 | UTR 3 | XP_016872355.1 |