Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
55 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616594 MIM: 604128 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ASAP3 - ArfGAP with SH3 domain, ankyrin repeat and PH domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143778.1 | 3469 | UTR 3 | NP_001137250.1 | |||
NM_017707.3 | 3469 | UTR 3 | NP_060177.2 | |||
XM_011541755.1 | 3469 | UTR 3 | XP_011540057.1 | |||
XM_017001685.1 | 3469 | UTR 3 | XP_016857174.1 | |||
XM_017001686.1 | 3469 | UTR 3 | XP_016857175.1 | |||
XM_017001687.1 | 3469 | UTR 3 | XP_016857176.1 | |||
XM_017001688.1 | 3469 | UTR 3 | XP_016857177.1 | |||
XM_017001689.1 | 3469 | UTR 3 | XP_016857178.1 |
TCEA3 - transcription elongation factor A3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |