Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
52 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171760 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALPL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ALPL - alkaline phosphatase, liver/bone/kidney | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000478.5 | Intron | NP_000469.3 | ||||
NM_001127501.3 | Intron | NP_001120973.2 | ||||
NM_001177520.2 | Intron | NP_001170991.1 | ||||
XM_005245818.1 | Intron | XP_005245875.1 | ||||
XM_006710546.2 | Intron | XP_006710609.1 | ||||
XM_017000903.1 | Intron | XP_016856392.1 |
Set Membership: |
HapMap |