Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607311 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PGR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
|||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
PGR - progesterone receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000926.4 | 12626 | UTR 3 | NP_000917.3 | |||
NM_001202474.3 | 12626 | UTR 3 | NP_001189403.1 | |||
NM_001271161.2 | 12626 | UTR 3 | NP_001258090.1 | |||
NM_001271162.1 | 12626 | UTR 3 | NP_001258091.1 | |||
XM_006718858.3 | 12626 | Intron | XP_006718921.1 | |||
XM_011542869.2 | 12626 | Intron | XP_011541171.1 |