Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGTACATTTTAACAGGGCTCCT[A/G]GAGAGGGCGGGGAGACGAAGCAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602332 MIM: 601664 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITPRIPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
ITPRIPL1 - inositol 1,4,5-trisphosphate receptor interacting protein-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008949.2 | 250 | UTR 5 | NP_001008949.1 | |||
NM_001163523.1 | 250 | Intron | NP_001156995.1 | |||
NM_001163524.1 | 250 | Intron | NP_001156996.1 | |||
NM_001324490.1 | 250 | Intron | NP_001311419.1 | |||
NM_178495.5 | 250 | Intron | NP_848590.3 | |||
XM_017003427.1 | 250 | UTR 5 | XP_016858916.1 |
LOC105373495 - uncharacterized LOC105373495 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCAPH - non-SMC condensin I complex subunit H | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNRNP200 - small nuclear ribonucleoprotein U5 subunit 200 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |