Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608860 MIM: 603068 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASTL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ASTL - astacin-like metallo-endopeptidase (M12 family) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002036.3 | 1204 | Intron | NP_001002036.3 | |||
XM_011511205.2 | 1204 | Intron | XP_011509507.1 | |||
XM_011511206.2 | 1204 | Intron | XP_011509508.1 | |||
XM_011511207.2 | 1204 | Intron | XP_011509509.1 | |||
XM_011511208.2 | 1204 | Intron | XP_011509510.1 |
DUSP2 - dual specificity phosphatase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004418.3 | 1204 | UTR 3 | NP_004409.1 | |||
XM_017003546.1 | 1204 | UTR 3 | XP_016859035.1 |
Set Membership: |
HapMap |