Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAGCGATTGCGTCTGGACTGAGT[C/T]CTGCCTGGGTGGGGTCAGGGCCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ASPG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASPG - asparaginase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080464.2 | Intron | NP_001073933.2 | ||||
XM_005267590.4 | Intron | XP_005267647.1 | ||||
XM_017021265.1 | Intron | XP_016876754.1 | ||||
XM_017021266.1 | Intron | XP_016876755.1 | ||||
XM_017021267.1 | Intron | XP_016876756.1 | ||||
XM_017021268.1 | Intron | XP_016876757.1 | ||||
XM_017021269.1 | Intron | XP_016876758.1 | ||||
XM_017021270.1 | Intron | XP_016876759.1 | ||||
XM_017021271.1 | Intron | XP_016876760.1 | ||||
XM_017021272.1 | Intron | XP_016876761.1 | ||||
XM_017021273.1 | Intron | XP_016876762.1 | ||||
XM_017021274.1 | Intron | XP_016876763.1 | ||||
XM_017021275.1 | Intron | XP_016876764.1 | ||||
XM_017021276.1 | Intron | XP_016876765.1 |
LOC105370691 - uncharacterized LOC105370691 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |