Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGGTCCCCAGCAGGCGAGGTCCC[C/T]AGCAGGCAAGTCTTCTGGAGGGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 111100 MIM: 136836 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FUT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FUT3 - fucosyltransferase 3 (Lewis blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000149.3 | 1880 | UTR 3 | NP_000140.1 | |||
NM_001097639.1 | 1880 | UTR 3 | NP_001091108.1 | |||
NM_001097640.1 | 1880 | UTR 3 | NP_001091109.1 | |||
NM_001097641.1 | 1880 | UTR 3 | NP_001091110.1 | |||
XM_011527865.1 | 1880 | UTR 3 | XP_011526167.1 | |||
XM_011527866.1 | 1880 | UTR 3 | XP_011526168.1 | |||
XM_011527867.1 | 1880 | UTR 3 | XP_011526169.1 |
FUT6 - fucosyltransferase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101928844 - uncharacterized LOC101928844 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |