Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 217050 | ||||||||||||||||||||
Literature Links: |
C6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C6 - complement component 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000065.3 | 3291 | UTR 3 | NP_000056.2 | |||
NM_001115131.2 | 3291 | UTR 3 | NP_001108603.2 | |||
XM_005248357.2 | 3291 | UTR 3 | XP_005248414.1 | |||
XM_006714496.3 | 3291 | UTR 3 | XP_006714559.1 | |||
XM_011514114.2 | 3291 | UTR 3 | XP_011512416.1 | |||
XM_011514115.2 | 3291 | UTR 3 | XP_011512417.1 | |||
XM_011514116.2 | 3291 | UTR 3 | XP_011512418.1 | |||
XM_011514117.2 | 3291 | UTR 3 | XP_011512419.1 | |||
XM_011514118.2 | 3291 | UTR 3 | XP_011512420.1 | |||
XM_011514119.2 | 3291 | UTR 3 | XP_011512421.1 | |||
XM_011514121.2 | 3291 | UTR 3 | XP_011512423.1 | |||
XM_017009818.1 | 3291 | UTR 3 | XP_016865307.1 | |||
XM_017009819.1 | 3291 | UTR 3 | XP_016865308.1 |