Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATTTGAAAGATTTCTTGGACCA[A/G]CGGCCACATCCCCCACAGGGCGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608460 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VPS9D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VPS9D1 - VPS9 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS9D1-AS1 - VPS9D1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF276 - zinc finger protein 276 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113525.1 | 80 | Intron | NP_001106997.1 | |||
NM_152287.3 | 80 | Intron | NP_689500.2 | |||
XM_005256324.3 | 80 | Intron | XP_005256381.1 | |||
XM_005256328.3 | 80 | Intron | XP_005256385.1 | |||
XM_017023889.1 | 80 | Intron | XP_016879378.1 | |||
XM_017023890.1 | 80 | UTR 5 | XP_016879379.1 |