Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613218 | ||||||||||||||||||||
Literature Links: |
CYB5A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYB5A - cytochrome b5 type A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190807.2 | 501 | Nonsense Mutation | TAA,TAC | *,Y 120 | NP_001177736.1 | |
NM_001914.3 | 501 | UTR 3 | NP_001905.1 | |||
NM_148923.3 | 501 | Nonsense Mutation | TAA,TAC | *,Y 130 | NP_683725.1 | |
XM_011525835.2 | 501 | Intron | XP_011524137.1 |