Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGGGAGGGCCTCCGAGTTCCCA[A/G]ACAAAACACAGGTACCTAAGGGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612395 MIM: 601987 MIM: 615775 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHKB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHKB - choline kinase beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CHKB-CPT1B - CHKB-CPT1B readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CPT1B - carnitine palmitoyltransferase 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145134.1 | 2736 | UTR 3 | NP_001138606.1 | |||
NM_001145135.1 | 2736 | UTR 3 | NP_001138607.1 | |||
NM_001145137.1 | 2736 | UTR 3 | NP_001138609.1 | |||
NM_004377.3 | 2736 | UTR 3 | NP_004368.1 | |||
NM_152245.2 | 2736 | UTR 3 | NP_689451.1 | |||
NM_152246.2 | 2736 | UTR 3 | NP_689452.1 |
SYCE3 - synaptonemal complex central element protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |