Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610473 | ||||||||||||||||||||
Literature Links: |
LPGAT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LPGAT1 - lysophosphatidylglycerol acyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320808.1 | 7651 | UTR 3 | NP_001307737.1 | |||
NM_014873.2 | 7651 | UTR 3 | NP_055688.1 | |||
XM_005273362.3 | 7651 | Intron | XP_005273419.1 | |||
XM_011510228.2 | 7651 | UTR 3 | XP_011508530.1 | |||
XM_011510229.2 | 7651 | UTR 3 | XP_011508531.1 | |||
XM_017003004.1 | 7651 | UTR 3 | XP_016858493.1 |