Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCTCACTTATATAAACACTGTA[C/T]TCCAAATTATTTTTTAATTTCCAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
42 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609287 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105378828 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105378828 - uncharacterized LOC105378828 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH3GLB1 - SH3 domain containing GRB2 like endophilin B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206651.1 | Intron | NP_001193580.1 | ||||
NM_001206652.1 | Intron | NP_001193581.1 | ||||
NM_001206653.1 | Intron | NP_001193582.1 | ||||
NM_016009.4 | Intron | NP_057093.1 | ||||
XM_006710672.2 | Intron | XP_006710735.1 | ||||
XM_006710673.2 | Intron | XP_006710736.1 | ||||
XM_011541538.1 | Intron | XP_011539840.1 | ||||
XM_017001408.1 | Intron | XP_016856897.1 | ||||
XM_017001409.1 | Intron | XP_016856898.1 | ||||
XM_017001410.1 | Intron | XP_016856899.1 |