Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGACAAGGGAGGGAGAAATGGAA[A/G]GAAGGAAAAGAGTTTAATAATTAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611587 | ||||||||||||||||||||
Literature Links: |
ARHGAP19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGAP19 - Rho GTPase activating protein 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204300.1 | 2179 | UTR 3 | NP_001191229.1 | |||
NM_001256423.1 | 2179 | UTR 3 | NP_001243352.1 | |||
NM_032900.5 | 2179 | UTR 3 | NP_116289.4 |
ARHGAP19-SLIT1 - ARHGAP19-SLIT1 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |