Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605458 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTR8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACTR8 - ARP8 actin-related protein 8 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022899.4 | 1662 | Intron | NP_075050.3 | |||
XM_005265587.4 | 1662 | Intron | XP_005265644.1 | |||
XM_011534249.2 | 1662 | Intron | XP_011532551.1 |
IL17RB - interleukin 17 receptor B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018725.3 | 1662 | UTR 3 | NP_061195.2 | |||
XM_005265310.4 | 1662 | UTR 3 | XP_005265367.1 | |||
XM_005265311.4 | 1662 | UTR 3 | XP_005265368.1 | |||
XM_005265312.4 | 1662 | UTR 3 | XP_005265369.1 | |||
XM_011533941.2 | 1662 | UTR 3 | XP_011532243.1 | |||
XM_017006804.1 | 1662 | UTR 3 | XP_016862293.1 | |||
XM_017006805.1 | 1662 | UTR 3 | XP_016862294.1 | |||
XM_017006806.1 | 1662 | UTR 3 | XP_016862295.1 | |||
XM_017006807.1 | 1662 | UTR 3 | XP_016862296.1 |
Set Membership: |
HapMap |