Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612377 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COMMD6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
COMMD6 - COMM domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287392.1 | 504 | UTR 3 | NP_001274321.1 | |||
NM_001287393.1 | 504 | UTR 3 | NP_001274322.1 | |||
NM_001287394.1 | 504 | UTR 3 | NP_001274323.1 | |||
NM_203495.3 | 504 | UTR 3 | NP_987091.1 | |||
NM_203497.3 | 504 | UTR 3 | NP_987093.1 | |||
XM_011534948.2 | 504 | UTR 3 | XP_011533250.1 |