Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171891 MIM: 613246 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDE1B - phosphodiesterase 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000924.3 | 1235 | Intron | NP_000915.1 | |||
NM_001165975.2 | 1235 | Intron | NP_001159447.1 | |||
NM_001288768.1 | 1235 | Intron | NP_001275697.1 | |||
NM_001288769.1 | 1235 | Intron | NP_001275698.1 | |||
NM_001315534.1 | 1235 | Intron | NP_001302463.1 | |||
NM_001315535.1 | 1235 | Intron | NP_001302464.1 | |||
XM_011538456.1 | 1235 | Intron | XP_011536758.1 | |||
XM_017019432.1 | 1235 | Intron | XP_016874921.1 | |||
XM_017019433.1 | 1235 | Intron | XP_016874922.1 |
PPP1R1A - protein phosphatase 1 regulatory inhibitor subunit 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006741.3 | 1235 | UTR 3 | NP_006732.3 | |||
XM_005268995.4 | 1235 | Intron | XP_005269052.1 | |||
XM_006719471.3 | 1235 | Intron | XP_006719534.1 |