Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604275 | ||||||||||||||||||||
Literature Links: |
CTNND2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTNND2 - catenin delta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288715.1 | 3913 | UTR 3 | NP_001275644.1 | |||
NM_001288716.1 | 3913 | UTR 3 | NP_001275645.1 | |||
NM_001288717.1 | 3913 | UTR 3 | NP_001275646.1 | |||
NM_001332.3 | 3913 | UTR 3 | NP_001323.1 | |||
XM_005248251.3 | 3913 | UTR 3 | XP_005248308.1 | |||
XM_005248252.2 | 3913 | UTR 3 | XP_005248309.1 | |||
XM_005248253.1 | 3913 | UTR 3 | XP_005248310.1 | |||
XM_011513967.2 | 3913 | UTR 3 | XP_011512269.1 | |||
XM_017009072.1 | 3913 | UTR 3 | XP_016864561.1 | |||
XM_017009073.1 | 3913 | UTR 3 | XP_016864562.1 | |||
XM_017009074.1 | 3913 | UTR 3 | XP_016864563.1 | |||
XM_017009075.1 | 3913 | UTR 3 | XP_016864564.1 | |||
XM_017009076.1 | 3913 | UTR 3 | XP_016864565.1 |