Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608466 MIM: 612684 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHSA1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AHSA1 - activator of Hsp90 ATPase activity 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ISM2 - isthmin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182509.3 | 2758 | UTR 3 | NP_872315.2 | |||
NM_199296.2 | 2758 | UTR 3 | NP_954993.1 | |||
XM_011536489.1 | 2758 | UTR 3 | XP_011534791.1 |