Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600924 MIM: 611256 MIM: 603927 MIM: 605915 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GFER PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GFER - growth factor, augmenter of liver regeneration | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005262.2 | 527 | Silent Mutation | CTG,TTG | L,L 153 | NP_005253.3 |
NOXO1 - NADPH oxidase organizer 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYNGR3 - synaptogyrin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBL3 - transducin beta like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |