Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191275 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCT PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DCT - dopachrome tautomerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129889.2 | 3144 | UTR 3 | NP_001123361.1 | |||
NM_001322182.1 | 3144 | UTR 3 | NP_001309111.1 | |||
NM_001322183.1 | 3144 | UTR 3 | NP_001309112.1 | |||
NM_001322184.1 | 3144 | UTR 3 | NP_001309113.1 | |||
NM_001322185.1 | 3144 | UTR 3 | NP_001309114.1 | |||
NM_001322186.1 | 3144 | UTR 3 | NP_001309115.1 | |||
NM_001922.4 | 3144 | UTR 3 | NP_001913.2 | |||
XM_011521049.1 | 3144 | UTR 3 | XP_011519351.1 | |||
XM_017020401.1 | 3144 | UTR 3 | XP_016875890.1 |
Set Membership: |
HapMap |