Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf39 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C14orf39 - chromosome 14 open reading frame 39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174978.2 | Intron | NP_777638.2 | ||||
XM_011536690.2 | Intron | XP_011534992.1 | ||||
XM_011536703.2 | Intron | XP_011535005.1 | ||||
XM_011536704.1 | Intron | XP_011535006.1 | ||||
XM_017021247.1 | Intron | XP_016876736.1 | ||||
XM_017021248.1 | Intron | XP_016876737.1 | ||||
XM_017021249.1 | Intron | XP_016876738.1 | ||||
XM_017021250.1 | Intron | XP_016876739.1 | ||||
XM_017021251.1 | Intron | XP_016876740.1 |