Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615168 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMZ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AMZ1 - archaelysin family metallopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284355.2 | Intron | NP_001271284.1 | ||||
NM_001321766.1 | Intron | NP_001308695.1 | ||||
NM_133463.3 | Intron | NP_597720.1 | ||||
XM_011515150.2 | Intron | XP_011513452.1 | ||||
XM_011515151.2 | Intron | XP_011513453.1 | ||||
XM_017011774.1 | Intron | XP_016867263.1 | ||||
XM_017011775.1 | Intron | XP_016867264.1 | ||||
XM_017011776.1 | Intron | XP_016867265.1 |
Set Membership: |
HapMap |