Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600428 | ||||||||||||||||||||
Literature Links: |
VAV2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VAV2 - vav guanine nucleotide exchange factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134398.1 | 4737 | UTR 3 | NP_001127870.1 | |||
NM_003371.3 | 4737 | UTR 3 | NP_003362.2 | |||
XM_005272213.1 | 4737 | UTR 3 | XP_005272270.1 | |||
XM_017015108.1 | 4737 | UTR 3 | XP_016870597.1 | |||
XM_017015109.1 | 4737 | UTR 3 | XP_016870598.1 | |||
XM_017015110.1 | 4737 | UTR 3 | XP_016870599.1 | |||
XM_017015111.1 | 4737 | UTR 3 | XP_016870600.1 | |||
XM_017015112.1 | 4737 | UTR 3 | XP_016870601.1 | |||
XM_017015113.1 | 4737 | Intron | XP_016870602.1 |