Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606525 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRGAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SRGAP3 - SLIT-ROBO Rho GTPase activating protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033117.2 | 6731 | UTR 3 | NP_001028289.1 | |||
NM_014850.3 | 6731 | UTR 3 | NP_055665.1 | |||
XM_011534295.1 | 6731 | UTR 3 | XP_011532597.1 | |||
XM_011534296.1 | 6731 | UTR 3 | XP_011532598.1 | |||
XM_011534301.2 | 6731 | UTR 3 | XP_011532603.1 | |||
XM_011534303.1 | 6731 | UTR 3 | XP_011532605.1 | |||
XM_017007574.1 | 6731 | UTR 3 | XP_016863063.1 | |||
XM_017007575.1 | 6731 | UTR 3 | XP_016863064.1 | |||
XM_017007576.1 | 6731 | UTR 3 | XP_016863065.1 | |||
XM_017007577.1 | 6731 | UTR 3 | XP_016863066.1 | |||
XM_017007578.1 | 6731 | UTR 3 | XP_016863067.1 | |||
XM_017007579.1 | 6731 | UTR 3 | XP_016863068.1 | |||
XM_017007580.1 | 6731 | Intron | XP_016863069.1 |
Set Membership: |
HapMap |