Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603699 | ||||||||||||||||||||
Literature Links: |
WNT11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WNT11 - Wnt family member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004626.2 | 1703 | UTR 3 | NP_004617.2 | |||
XM_005274231.1 | 1703 | UTR 3 | XP_005274288.1 | |||
XM_011545238.1 | 1703 | UTR 3 | XP_011543540.1 | |||
XM_011545239.2 | 1703 | UTR 3 | XP_011543541.1 | |||
XM_011545240.2 | 1703 | UTR 3 | XP_011543542.1 | |||
XM_011545241.2 | 1703 | UTR 3 | XP_011543543.1 | |||
XM_017018241.1 | 1703 | UTR 3 | XP_016873730.1 | |||
XM_017018242.1 | 1703 | UTR 3 | XP_016873731.1 | |||
XM_017018243.1 | 1703 | UTR 3 | XP_016873732.1 |