Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCAGGAGTTTGAGATCAGCCTC[A/G]GAAACACAGTAGAACCCCATCTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614363 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSBG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACSBG2 - acyl-CoA synthetase bubblegum family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289177.1 | Intron | NP_001276106.1 | ||||
NM_001289178.1 | Intron | NP_001276107.1 | ||||
NM_001289179.1 | Intron | NP_001276108.1 | ||||
NM_001289180.1 | Intron | NP_001276109.1 | ||||
NM_001321384.1 | Intron | NP_001308313.1 | ||||
NM_030924.4 | Intron | NP_112186.3 | ||||
XM_005259653.3 | Intron | XP_005259710.1 | ||||
XM_011528329.1 | Intron | XP_011526631.1 | ||||
XM_017027332.1 | Intron | XP_016882821.1 | ||||
XM_017027333.1 | Intron | XP_016882822.1 | ||||
XM_017027334.1 | Intron | XP_016882823.1 |
LOC105372255 - uncharacterized LOC105372255 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |