Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACACCCAATTCTCCAAGAGGGTCT[C/G]TTGGACTTTCTCTCACTTGCCCGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614363 | ||||||||||||||||||||
Literature Links: |
ACSBG2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACSBG2 - acyl-CoA synthetase bubblegum family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289177.1 | Intron | NP_001276106.1 | ||||
NM_001289178.1 | Intron | NP_001276107.1 | ||||
NM_001289179.1 | Intron | NP_001276108.1 | ||||
NM_001289180.1 | Intron | NP_001276109.1 | ||||
NM_001321384.1 | Intron | NP_001308313.1 | ||||
NM_030924.4 | Intron | NP_112186.3 | ||||
XM_005259653.3 | Intron | XP_005259710.1 | ||||
XM_011528329.1 | Intron | XP_011526631.1 | ||||
XM_017027332.1 | Intron | XP_016882821.1 | ||||
XM_017027333.1 | Intron | XP_016882822.1 | ||||
XM_017027334.1 | Intron | XP_016882823.1 |
LOC105372255 - uncharacterized LOC105372255 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |