Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAAAATTTCTGGGCATCTTAAG[C/G]TTTATATCAATCAGGGACTACCTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609488 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf47 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C4orf47 - chromosome 4 open reading frame 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114357.1 | Intron | NP_001107829.1 | ||||
XM_005263019.3 | Intron | XP_005263076.1 | ||||
XM_011531989.2 | Intron | XP_011530291.1 | ||||
XM_017008236.1 | Intron | XP_016863725.1 | ||||
XM_017008237.1 | Intron | XP_016863726.1 | ||||
XM_017008238.1 | Intron | XP_016863727.1 | ||||
XM_017008239.1 | Intron | XP_016863728.1 | ||||
XM_017008240.1 | Intron | XP_016863729.1 | ||||
XM_017008241.1 | Intron | XP_016863730.1 | ||||
XM_017008242.1 | Intron | XP_016863731.1 |
CCDC110 - coiled-coil domain containing 110 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145411.1 | Intron | NP_001138883.1 | ||||
NM_152775.3 | Intron | NP_689988.1 | ||||
XM_005262891.3 | Intron | XP_005262948.1 | ||||
XM_006714172.2 | Intron | XP_006714235.1 | ||||
XM_011531827.2 | Intron | XP_011530129.1 | ||||
XM_011531828.2 | Intron | XP_011530130.1 | ||||
XM_011531829.2 | Intron | XP_011530131.1 | ||||
XM_017007987.1 | Intron | XP_016863476.1 | ||||
XM_017007988.1 | Intron | XP_016863477.1 |
Set Membership: |
HapMap Validated |