Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605501 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPHOSPH9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MPHOSPH9 - M-phase phosphoprotein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022782.3 | 3965 | UTR 3 | NP_073619.3 | |||
XM_011537740.2 | 3965 | Intron | XP_011536042.1 | |||
XM_011537741.2 | 3965 | Intron | XP_011536043.1 | |||
XM_017018673.1 | 3965 | UTR 3 | XP_016874162.1 | |||
XM_017018674.1 | 3965 | UTR 3 | XP_016874163.1 | |||
XM_017018675.1 | 3965 | UTR 3 | XP_016874164.1 | |||
XM_017018676.1 | 3965 | UTR 3 | XP_016874165.1 | |||
XM_017018677.1 | 3965 | UTR 3 | XP_016874166.1 | |||
XM_017018678.1 | 3965 | UTR 3 | XP_016874167.1 | |||
XM_017018679.1 | 3965 | UTR 3 | XP_016874168.1 | |||
XM_017018680.1 | 3965 | UTR 3 | XP_016874169.1 | |||
XM_017018681.1 | 3965 | UTR 3 | XP_016874170.1 |