Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109135 MIM: 605800 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AXL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AXL - AXL receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HNRNPUL1 - heterogeneous nuclear ribonucleoprotein U like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301016.2 | Intron | NP_001287945.1 | ||||
NM_001321208.1 | Intron | NP_001308137.1 | ||||
NM_001321211.1 | Intron | NP_001308140.1 | ||||
NM_007040.5 | Intron | NP_008971.2 | ||||
NM_144732.4 | Intron | NP_653333.1 | ||||
XM_005258459.3 | Intron | XP_005258516.1 | ||||
XM_005258461.4 | Intron | XP_005258518.1 | ||||
XM_005258463.3 | Intron | XP_005258520.1 | ||||
XM_005258464.3 | Intron | XP_005258521.1 | ||||
XM_005258465.4 | Intron | XP_005258522.1 | ||||
XM_011526392.2 | Intron | XP_011524694.1 | ||||
XM_011526393.2 | Intron | XP_011524695.1 | ||||
XM_011526394.2 | Intron | XP_011524696.1 | ||||
XM_011526395.2 | Intron | XP_011524697.1 |