Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603675 MIM: 602102 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC723805 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC723805 - interleukin-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS16 - ribosomal protein S16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SUPT5H - SPT5 homolog, DSIF elongation factor subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111020.2 | 215 | UTR 5 | NP_001104490.1 | |||
NM_001130824.1 | 215 | Intron | NP_001124296.1 | |||
NM_001130825.1 | 215 | UTR 5 | NP_001124297.1 | |||
NM_001319990.1 | 215 | Intron | NP_001306919.1 | |||
NM_001319991.1 | 215 | Intron | NP_001306920.1 | |||
NM_003169.3 | 215 | UTR 5 | NP_003160.2 | |||
XM_017027174.1 | 215 | Intron | XP_016882663.1 |