Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGCTTGTGCTTGAAGGTGGTCAG[C/T]TTTTTCTTTAGGAGATAAGGAAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608153 MIM: 605124 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R14A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPP1R14A - protein phosphatase 1 regulatory inhibitor subunit 14A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243947.1 | Intron | NP_001230876.1 | ||||
NM_033256.2 | Intron | NP_150281.1 |
SPINT2 - serine peptidase inhibitor, Kunitz type 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |