Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164731 MIM: 600137 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AKT2 - AKT serine/threonine kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CNTD2 - cyclin N-terminal domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024877.3 | 1515 | UTR 3 | NP_079153.2 | |||
XM_006723395.3 | 1515 | Intron | XP_006723458.2 |
MAP3K10 - mitogen-activated protein kinase kinase kinase 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC9B - tetratricopeptide repeat domain 9B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |