Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCAGATAGCACCTGATGTTGGCC[A/G]TTATGACAGAAATTAAATGGTCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608383 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPYSL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DPYSL5 - dihydropyrimidinase like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253723.1 | Intron | NP_001240652.1 | ||||
NM_001253724.1 | Intron | NP_001240653.1 | ||||
NM_020134.3 | Intron | NP_064519.2 |
Set Membership: |
HapMap Validated |