Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604626 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ME3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ME3 - malic enzyme 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014811.1 | 1816 | Silent Mutation | NP_001014811.1 | |||
NM_001161586.1 | 1816 | Silent Mutation | NP_001155058.1 | |||
NM_006680.2 | 1816 | Silent Mutation | NP_006671.2 | |||
XM_005273717.1 | 1816 | Silent Mutation | XP_005273774.1 | |||
XM_017017136.1 | 1816 | Silent Mutation | XP_016872625.1 | |||
XM_017017137.1 | 1816 | Silent Mutation | XP_016872626.1 | |||
XM_017017138.1 | 1816 | Silent Mutation | XP_016872627.1 |
Set Membership: |
HapMap Validated |