Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATCATAGGTAAAGCCCTTGGCA[C/T]ATAACCTAGTTCTTGGAAGACTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR107 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR107 - G protein-coupled receptor 107 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136557.1 | Intron | NP_001130029.1 | ||||
NM_001136558.1 | Intron | NP_001130030.1 | ||||
NM_001287346.1 | Intron | NP_001274275.1 | ||||
NM_020960.4 | Intron | NP_066011.2 | ||||
XM_017014953.1 | Intron | XP_016870442.1 | ||||
XM_017014954.1 | Intron | XP_016870443.1 |