Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162641 MIM: 602001 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPY1R PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NPY1R - neuropeptide Y receptor Y1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000909.5 | 230 | Intron | NP_000900.1 | |||
XM_005263031.3 | 230 | Intron | XP_005263088.1 | |||
XM_011532010.2 | 230 | Intron | XP_011530312.1 |
NPY5R - neuropeptide Y receptor Y5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317091.1 | 230 | Intron | NP_001304020.1 | |||
NM_001317092.1 | 230 | Intron | NP_001304021.1 | |||
NM_006174.3 | 230 | Intron | NP_006165.1 | |||
XM_005263038.3 | 230 | UTR 5 | XP_005263095.1 | |||
XM_011532015.2 | 230 | UTR 5 | XP_011530317.1 | |||
XM_011532017.2 | 230 | UTR 5 | XP_011530319.1 | |||
XM_017008255.1 | 230 | Intron | XP_016863744.1 | |||
XM_017008256.1 | 230 | UTR 5 | XP_016863745.1 |