Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610576 MIM: 156560 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGAP9 - Rho GTPase activating protein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080156.2 | Intron | NP_001073625.1 | ||||
NM_001080157.1 | Intron | NP_001073626.1 | ||||
NM_001319850.1 | Intron | NP_001306779.1 | ||||
NM_001319851.1 | Intron | NP_001306780.1 | ||||
NM_001319852.1 | Intron | NP_001306781.1 | ||||
NM_032496.3 | Intron | NP_115885.2 | ||||
XM_005269083.2 | Intron | XP_005269140.1 | ||||
XM_005269084.2 | Intron | XP_005269141.1 | ||||
XM_005269085.2 | Intron | XP_005269142.1 | ||||
XM_011538656.2 | Intron | XP_011536958.1 | ||||
XM_011538657.2 | Intron | XP_011536959.1 | ||||
XM_011538658.2 | Intron | XP_011536960.1 | ||||
XM_011538659.2 | Intron | XP_011536961.1 | ||||
XM_017019800.1 | Intron | XP_016875289.1 | ||||
XM_017019801.1 | Intron | XP_016875290.1 | ||||
XM_017019802.1 | Intron | XP_016875291.1 | ||||
XM_017019803.1 | Intron | XP_016875292.1 | ||||
XM_017019804.1 | Intron | XP_016875293.1 |
MARS - methionyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004990.3 | Intron | NP_004981.2 | ||||
XM_006719398.3 | Intron | XP_006719461.1 |
Set Membership: |
HapMap |