Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTCCCTTTCTACTTATCTGCTA[A/C]AGAAGTAATTTTTAAAAGTGGATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C15orf57 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C15orf57 - chromosome 15 open reading frame 57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080791.2 | Intron | NP_001074260.1 | ||||
NM_001080792.2 | Intron | NP_001074261.1 | ||||
NM_001289132.1 | Intron | NP_001276061.1 | ||||
NM_052849.3 | Intron | NP_443081.1 |
Set Membership: |
HapMap |