Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTCACAAGGTCTAGGTCAGCCA[C/T]AGCAGGCCTGTGCAGGGAAGGCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616476 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGBL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AGBL4 - ATP/GTP binding protein like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323573.1 | 1820 | Intron | NP_001310502.1 | |||
NM_001323574.1 | 1820 | Intron | NP_001310503.1 | |||
NM_001323575.1 | 1820 | Intron | NP_001310504.1 | |||
NM_032785.3 | 1820 | Intron | NP_116174.3 | |||
XM_011542308.2 | 1820 | Intron | XP_011540610.1 | |||
XM_011542310.2 | 1820 | Intron | XP_011540612.1 | |||
XM_017002595.1 | 1820 | UTR 3 | XP_016858084.1 | |||
XM_017002596.1 | 1820 | Intron | XP_016858085.1 | |||
XM_017002597.1 | 1820 | Intron | XP_016858086.1 | |||
XM_017002598.1 | 1820 | Intron | XP_016858087.1 |
Set Membership: |
HapMap |