Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609010 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCCC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MCCC1 - methylcrotonoyl-CoA carboxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293273.1 | Intron | NP_001280202.1 | ||||
NM_020166.4 | Intron | NP_064551.3 | ||||
XM_006713702.1 | Intron | XP_006713765.1 | ||||
XM_011512992.2 | Intron | XP_011511294.1 | ||||
XM_011512993.1 | Intron | XP_011511295.1 | ||||
XM_017006874.1 | Intron | XP_016862363.1 | ||||
XM_017006875.1 | Intron | XP_016862364.1 |
Set Membership: |
HapMap |