Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610106 MIM: 612931 MIM: 606344 | ||||||||||||||||||||
Literature Links: |
DBNL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DBNL - drebrin like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014436.2 | 737 | Intron | NP_001014436.1 | |||
NM_001122956.1 | 737 | Intron | NP_001116428.1 | |||
NM_001284313.1 | 737 | Intron | NP_001271242.1 | |||
NM_001284315.1 | 737 | Intron | NP_001271244.1 | |||
NM_014063.6 | 737 | Intron | NP_054782.2 |
PGAM2 - phosphoglycerate mutase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000290.3 | 737 | Missense Mutation | ACC,CCC | T,P 227 | NP_000281.2 |
POLM - DNA polymerase mu | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |