Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603961 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEMA3A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SEMA3A - semaphorin 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006080.2 | 6667 | Intron | NP_006071.1 | |||
XM_005250110.3 | 6667 | UTR 3 | XP_005250167.1 | |||
XM_005250111.4 | 6667 | UTR 3 | XP_005250168.1 | |||
XM_006715839.3 | 6667 | UTR 3 | XP_006715902.1 | |||
XM_011515734.2 | 6667 | UTR 3 | XP_011514036.1 | |||
XM_017011673.1 | 6667 | UTR 3 | XP_016867162.1 |
Set Membership: |
HapMap |