Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
64 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608661 MIM: 604471 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PODN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PODN - podocan | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC1A7 - solute carrier family 1 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287595.1 | 1525 | Silent Mutation | AGC,AGT | S,S 613 | NP_001274524.1 | |
NM_001287596.1 | 1525 | Intron | NP_001274525.1 | |||
NM_001287597.1 | 1525 | UTR 3 | NP_001274526.1 | |||
NM_006671.5 | 1525 | Silent Mutation | AGC,AGT | S,S 554 | NP_006662.3 | |
XM_011542002.2 | 1525 | Silent Mutation | AGC,AGT | S,S 331 | XP_011540304.1 |
Set Membership: |
HapMap |