Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 616365 | |||||||||||||||||||||||
Literature Links: |
DEPDC4 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
DEPDC4 - DEP domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCYL2 - SCY1 like pseudokinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317784.1 | Intron | NP_001304713.1 | ||||
NM_017988.5 | Intron | NP_060458.3 | ||||
XM_017019618.1 | Intron | XP_016875107.1 | ||||
XM_017019619.1 | Intron | XP_016875108.1 | ||||
XM_017019620.1 | Intron | XP_016875109.1 |
Set Membership: |
HapMap |