Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605262 MIM: 603398 | ||||||||||||||||||||
Literature Links: |
NDRG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NDRG1 - N-myc downstream regulated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135242.1 | 2316 | UTR 3 | NP_001128714.1 | |||
NM_001258432.1 | 2316 | UTR 3 | NP_001245361.1 | |||
NM_001258433.1 | 2316 | UTR 3 | NP_001245362.1 | |||
NM_006096.3 | 2316 | UTR 3 | NP_006087.2 | |||
XM_011516791.1 | 2316 | UTR 3 | XP_011515093.1 | |||
XM_011516792.1 | 2316 | UTR 3 | XP_011515094.1 |
WISP1 - WNT1 inducible signaling pathway protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |