Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
39 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614453 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LRRC7 - leucine rich repeat containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020794.2 | Intron | NP_065845.1 | ||||
XM_017001885.1 | Intron | XP_016857374.1 | ||||
XM_017001886.1 | Intron | XP_016857375.1 | ||||
XM_017001887.1 | Intron | XP_016857376.1 | ||||
XM_017001888.1 | Intron | XP_016857377.1 | ||||
XM_017001889.1 | Intron | XP_016857378.1 | ||||
XM_017001890.1 | Intron | XP_016857379.1 | ||||
XM_017001891.1 | Intron | XP_016857380.1 | ||||
XM_017001892.1 | Intron | XP_016857381.1 | ||||
XM_017001893.1 | Intron | XP_016857382.1 | ||||
XM_017001894.1 | Intron | XP_016857383.1 | ||||
XM_017001895.1 | Intron | XP_016857384.1 |
Set Membership: |
HapMap |